Assay Details
| Gene Symbol: SMYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204155.1 | NP_989486.1 | 1 - 2 | 118 | 66 | |
| XM_015285868.2 | XP_015141354.1 | 1 - 2 | 402 | 66 | ||
| GenBank mRNA | AF410781.1 | - | 1 - 2 | 118 | 66 | |
| AF410782.1 | - | 1 - 2 | 118 | 66 | ||
More Information
Set Membership: |
|

SMYD1
Genomic Map