Assay Details
| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204170.2 | NP_989501.1 | 1 - 2 | 225 | 61 | |
| GenBank mRNA | AB053163.1 | - | 1 - 2 | 227 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCNA| Gene Symbol: PCNA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204170.2 | NP_989501.1 | 1 - 2 | 225 | 61 | |
| GenBank mRNA | AB053163.1 | - | 1 - 2 | 227 | 61 | |
Set Membership: |
|