Assay Details
| Gene Symbol: TPD52L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204215.1 | NP_989546.1 | 6 - 7 | 445 | 83 | |
| XM_015284430.2 | XP_015139916.1 | 5 - 6 | 483 | 83 | ||
| XM_015284431.2 | XP_015139917.1 | 7 - 8 | 578 | 83 | ||
| GenBank mRNA | AY004870.1 | - | 6 - 7 | 445 | 83 | |
More Information
Set Membership: |
|

TPD52L1
Genomic Map