Assay Details
| Gene Symbol: PKNOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204226.1 | NP_989557.1 | 2 - 3 | 217 | 56 | |
| XM_015297933.2 | XP_015153419.1 | 7 - 8 | 768 | 56 | ||
| XM_015297934.2 | XP_015153420.1 | 6 - 7 | 699 | 56 | ||
| XM_015297935.2 | XP_015153421.1 | 7 - 8 | 686 | 56 | ||
| XM_015297936.2 | XP_015153422.1 | 8 - 9 | 1303 | 56 | ||
| GenBank mRNA | AB086816.1 | - | 2 - 3 | 217 | 56 | |
More Information
Set Membership: |
|

PKNOX2
Genomic Map