Assay Details
| Gene Symbol: PDGFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204306.1 | NP_989637.1 | 3 - 4 | 412 | 65 | |
| GenBank mRNA | AB031021.1 | - | 3 - 4 | 321 | 65 | |
| AB031022.1 | - | 3 - 4 | 633 | 65 | ||
| AB031023.1 | - | 3 - 4 | 412 | 65 | ||
| AF188841.1 | - | 3 - 4 | 266 | 65 | ||
| BX950584.1 | - | 3 - 4 | 286 | 65 | ||
More Information
Set Membership: |
|

PDGFA
Genomic Map