Assay Details
| Gene Symbol: ACVR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204317.1 | NP_989648.1 | 10 - 11 | 1610 | 74 | |
| GenBank mRNA | KX714223.1 | - | 10 - 11 | 1346 | 74 | |
| U31223.1 | - | 10 - 11 | 1610 | 74 | ||
More Information
Set Membership: |
|

ACVR2B
Genomic Map