Assay Details
| Gene Symbol: FOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204328.1 | NP_989659.1 | 1 - 2 | 861 | 62 | |
| GenBank mRNA | AF114261.1 | - | 1 - 2 | 861 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXO1| Gene Symbol: FOXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204328.1 | NP_989659.1 | 1 - 2 | 861 | 62 | |
| GenBank mRNA | AF114261.1 | - | 1 - 2 | 861 | 62 | |
Set Membership: |
|