Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204357.1 | NP_989688.1 | 10 - 11 | 1024 | 97 | |
| XM_015287267.2 | XP_015142753.1 | 11 - 12 | 1211 | 97 | ||
| XM_015287269.2 | XP_015142755.1 | 8 - 9 | 843 | 97 | ||
| GenBank mRNA | AF085247.1 | - | 10 - 11 | 1024 | 97 | |
| BX929476.2 | - | 8 - 9 | 859 | 97 | ||
More Information
Set Membership: |
|

ATXN3
Genomic Map