Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204357.1 | NP_989688.1 | 3 - 4 | 269 | 103 | |
| XM_015287267.2 | XP_015142753.1 | 3 - 4 | 420 | 103 | ||
| XM_015287268.2 | XP_015142754.1 | 3 - 4 | 271 | 103 | ||
| GenBank mRNA | AF085247.1 | - | 3 - 4 | 269 | 103 | |
More Information
Set Membership: |
|

ATXN3
Genomic Map