Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204357.1 | NP_989688.1 | 4 - 5 | 347 | 141 | |
| XM_015287267.2 | XP_015142753.1 | 4 - 5 | 498 | 177 | ||
| XM_015287268.2 | XP_015142754.1 | 4 - 5 | 349 | 177 | ||
| GenBank mRNA | AF085247.1 | - | 4 - 5 | 347 | 141 | |
More Information
Set Membership: |
|

ATXN3
Genomic Map