Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204357.1 | NP_989688.1 | 5 - 6 | 414 | 116 | |
| GenBank mRNA | AF085247.1 | - | 5 - 6 | 414 | 116 | |
More Information
Set Membership: |
|

ATXN3
Genomic Map