Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204357.1 | NP_989688.1 | 6 - 7 | 509 | 67 | |
| XM_015287267.2 | XP_015142753.1 | 7 - 8 | 696 | 67 | ||
| XM_015287268.2 | XP_015142754.1 | 7 - 8 | 547 | 67 | ||
| XM_015287269.2 | XP_015142755.1 | 4 - 5 | 328 | 67 | ||
| GenBank mRNA | AF085247.1 | - | 6 - 7 | 509 | 67 | |
| BX929476.2 | - | 4 - 5 | 344 | 67 | ||
| CR353700.1 | - | 3 - 4 | 319 | 67 | ||
More Information
Set Membership: |
|

ATXN3
Genomic Map