Assay Details
| Gene Symbol: ATXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204357.1 | NP_989688.1 | 9 - 10 | 941 | 113 | |
| XM_015287267.2 | XP_015142753.1 | 10 - 11 | 113 | |||
| XM_015287269.2 | XP_015142755.1 | 7 - 8 | 113 | |||
| GenBank mRNA | AF085247.1 | - | 9 - 10 | 941 | 113 | |
| BX929476.2 | - | 7 - 8 | 776 | 113 | ||
More Information
Set Membership: |
|

ATXN3
Genomic Map