Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204385.1 | NP_989716.1 | 2 - 3 | 246 | 74 | |
| XM_015290232.2 | XP_015145718.1 | 2 - 3 | 74 | |||
| XM_025152733.1 | XP_025008501.1 | 3 - 4 | 74 | |||
| XM_025152734.1 | XP_025008502.1 | 2 - 3 | 74 | |||
| GenBank mRNA | AY297457.1 | - | 2 - 3 | 246 | 74 | |
| BX929559.2 | - | 1 - 2 | 74 | |||
| BX950639.2 | - | 2 - 3 | 246 | 74 | ||
More Information
Set Membership: |
|

RGS4
Genomic Map