Assay Details
| Gene Symbol: ATP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204418.1 | NP_989749.1 | 2 - 3 | 395 | 69 | |
| GenBank mRNA | L08047.1 | - | 2 - 3 | 395 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP4B| Gene Symbol: ATP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204418.1 | NP_989749.1 | 2 - 3 | 395 | 69 | |
| GenBank mRNA | L08047.1 | - | 2 - 3 | 395 | 69 | |
Set Membership: |
|