Assay Details
| Gene Symbol: OCM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204422.1 | NP_989753.1 | 4 - 5 | 409 | 133 | |
| XM_015294127.2 | XP_015149613.1 | 4 - 5 | 133 | |||
| GenBank mRNA | U03850.1 | - | 4 - 5 | 409 | 133 | |
More Information
Set Membership: |
|

OCM
Genomic Map