Assay Details
| Gene Symbol: TERF2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204468.1 | NP_989799.1 | 1 - 2 | 496 | 123 | |
| GenBank mRNA | AY083608.1 | - | 1 - 2 | 496 | 123 | |
More Information
Set Membership: |
|

TERF2IP
Genomic Map