Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204469.1 | NP_989800.1 | 5 - 6 | 789 | 60 | |
| GenBank mRNA | AY339892.1 | - | 5 - 6 | 789 | 60 | |
| BX950349.1 | - | 5 - 6 | 770 | 60 | ||
More Information
Set Membership: |
|

AIP
Genomic Map