Assay Details
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205097.1 | NP_990428.1 | 7 - 8 | 1436 | 51 | |
| XM_015286981.2 | XP_015142467.1 | 8 - 9 | 7248 | 51 | ||
| XM_015286983.2 | XP_015142469.1 | 8 - 9 | 7089 | 51 | ||
| XM_015286984.2 | XP_015142470.1 | 7 - 8 | 7071 | 51 | ||
| XM_015286985.2 | XP_015142471.1 | 7 - 8 | 7059 | 51 | ||
| XM_015286986.2 | XP_015142472.1 | 6 - 7 | 6879 | 51 | ||
| GenBank mRNA | AF000138.1 | - | 7 - 8 | 1436 | 51 | |
| CR386643.1 | - | 7 - 8 | 1290 | 51 | ||
More Information
Set Membership: |
|

RAPSN
Genomic Map