Assay Details
| Gene Symbol: ZEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205131.1 | NP_990462.1 | 4 - 5 | 497 | 66 | |
| XM_015281304.2 | XP_015136790.1 | 4 - 5 | 552 | 66 | ||
| XM_015281305.1 | XP_015136791.1 | 4 - 5 | 538 | 66 | ||
| XM_025147055.1 | XP_025002823.1 | 5 - 6 | 651 | 66 | ||
| XM_025147056.1 | XP_025002824.1 | 4 - 5 | 561 | 66 | ||
| GenBank mRNA | D14313.1 | - | 4 - 5 | 497 | 66 | |
| KY362200.1 | - | 4 - 5 | 486 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015281305.1, XM_025147056.1 transcripts.
Set Membership: |
|

ZEB1
Genomic Map