Assay Details
| Gene Symbol: NR2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205170.1 | NP_990501.1 | 8 - 9 | 1322 | 61 | |
| GenBank mRNA | S72373.1 | - | 8 - 9 | 1322 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NR2E1| Gene Symbol: NR2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205170.1 | NP_990501.1 | 8 - 9 | 1322 | 61 | |
| GenBank mRNA | S72373.1 | - | 8 - 9 | 1322 | 61 | |
Set Membership: |
|