Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205255.1 | NP_990586.1 | 3 - 4 | 464 | 65 | |
| XM_015284952.2 | XP_015140438.1 | 3 - 4 | 515 | 65 | ||
| XM_015284953.2 | XP_015140439.1 | 3 - 4 | 504 | 65 | ||
| XM_015284954.2 | XP_015140440.1 | 3 - 4 | 498 | 65 | ||
| XM_015284955.2 | XP_015140441.1 | 3 - 4 | 579 | 65 | ||
| XM_015284956.2 | XP_015140442.1 | 3 - 4 | 5386 | 65 | ||
| XM_025148556.1 | XP_025004324.1 | 4 - 5 | 638 | 65 | ||
| XM_025148557.1 | XP_025004325.1 | 3 - 4 | 466 | 65 | ||
| XM_025148558.1 | XP_025004326.1 | 3 - 4 | 820 | 65 | ||
| XM_025148559.1 | XP_025004327.1 | 3 - 4 | 540 | 65 | ||
| XM_025148560.1 | XP_025004328.1 | 3 - 4 | 484 | 65 | ||
| XM_025148561.1 | XP_025004329.1 | 3 - 4 | 393 | 65 | ||
| GenBank mRNA | BX933939.1 | - | 3 - 4 | 487 | 65 | |
| X63530.1 | - | 464 | 65 | |||
More Information
Set Membership: |
|

VSNL1
Genomic Map