Assay Details
| Gene Symbol: AKTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005838.1 | NP_001005838.1 | 2 - 3 | 190 | 66 | |
| XM_015292357.2 | XP_015147843.1 | 2 - 3 | 271 | 66 | ||
| XM_015292358.2 | XP_015147844.1 | 2 - 3 | 263 | 66 | ||
| XM_015292359.2 | XP_015147845.1 | 2 - 3 | 214 | 66 | ||
| XM_025154159.1 | XP_025009927.1 | 2 - 3 | 151 | 66 | ||
| GenBank mRNA | AJ720439.1 | - | 2 - 3 | 190 | 66 | |
More Information
Set Membership: |
|

AKTIP
Genomic Map