Assay Details
| Gene Symbol: USP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006130.1 | NP_001006130.1 | 5 - 6 | 1337 | 60 | |
| XM_025154184.1 | XP_025009952.1 | 5 - 6 | 1315 | 60 | ||
| GenBank mRNA | AJ720400.1 | - | 5 - 6 | 1337 | 60 | |
More Information
Set Membership: |
|

USP10
Genomic Map