Assay Details
| Gene Symbol: METTL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006172.2 | NP_001006172.1 | 3 - 4 | 688 | 96 | |
| XM_015294230.2 | XP_015149716.1 | 3 - 4 | 682 | 96 | ||
| GenBank mRNA | AJ719408.1 | - | 3 - 4 | 690 | 96 | |
More Information
Set Membership: |
|

METTL9
Genomic Map