Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006253.1 | NP_001006253.1 | 3 - 4 | 377 | 64 | |
| XM_025149296.1 | XP_025005064.1 | 4 - 5 | 2964 | 64 | ||
| GenBank mRNA | AJ719633.1 | - | 3 - 4 | 377 | 64 | |
More Information
Set Membership: |
|

PCNP
Genomic Map