Assay Details
| Gene Symbol: AP1S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006261.1 | NP_001006261.1 | 2 - 3 | 353 | 72 | |
| XM_015273167.2 | XP_015128653.1 | 2 - 3 | 273 | 72 | ||
| XM_015273183.2 | XP_015128669.1 | 2 - 3 | 342 | 72 | ||
| XM_015273190.1 | XP_015128676.1 | 2 - 3 | 361 | 72 | ||
| XM_025149728.1 | XP_025005496.1 | 2 - 3 | 365 | 72 | ||
| XM_025149733.1 | XP_025005501.1 | 2 - 3 | 804 | 72 | ||
| XM_025149735.1 | XP_025005503.1 | 2 - 3 | 835 | 72 | ||
| XM_025149737.1 | XP_025005505.1 | 2 - 3 | 304 | 72 | ||
| XM_025149740.1 | XP_025005508.1 | 2 - 3 | 761 | 72 | ||
| XM_025149742.1 | XP_025005510.1 | 2 - 3 | 287 | 72 | ||
| XM_025149747.1 | XP_025005515.1 | 2 - 3 | 387 | 72 | ||
| GenBank mRNA | AJ720066.1 | - | 2 - 3 | 353 | 72 | |
| AJ720312.1 | - | 2 - 3 | 334 | 72 | ||
More Information
Set Membership: |
|

AP1S2
Genomic Map