Assay Details
| Gene Symbol: EEF1AKMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006277.1 | NP_001006277.1 | 2 - 3 | 189 | 101 | |
| XM_015278046.2 | XP_015133532.1 | 2 - 3 | 304 | 101 | ||
| GenBank mRNA | AJ719998.1 | - | 2 - 3 | 189 | 101 | |
More Information
Set Membership: |
|

EEF1AKMT1
Genomic Map