Assay Details
| Gene Symbol: EEF1AKMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006277.1 | NP_001006277.1 | 3 - 4 | 271 | 71 | |
| XM_015278046.2 | XP_015133532.1 | 3 - 4 | 386 | 71 | ||
| GenBank mRNA | AJ719998.1 | - | 3 - 4 | 271 | 71 | |
More Information
Set Membership: |
|

EEF1AKMT1
Genomic Map