Assay Details
| Gene Symbol: ADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006290.2 | NP_001006290.1 | 2 - 3 | 123 | 61 | |
| GenBank mRNA | AJ720038.1 | - | 2 - 3 | 124 | 61 | |
More Information
Set Membership: |
|

ADA
Genomic Map