Assay Details
| Gene Symbol: HMGN2P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006412.1 | NP_001006412.1 | 4 - 5 | 219 | 71 | |
| XM_015284632.2 | XP_015140118.1 | 4 - 5 | 249 | 71 | ||
| GenBank mRNA | AJ720719.1 | - | 4 - 5 | 219 | 71 | |
More Information
Set Membership: |
|

HMGN2P46
Genomic Map