Assay Details
| Gene Symbol: DCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006451.1 | NP_001006451.1 | 2 - 3 | 253 | 69 | |
| GenBank mRNA | AJ719431.1 | - | 2 - 3 | 253 | 69 | |
| AJ719977.1 | - | 2 - 3 | 297 | 69 | ||
| EU835757.1 | - | 2 - 3 | 185 | 69 | ||
More Information
Set Membership: |
|

DCK
Genomic Map