Assay Details
| Gene Symbol: NHLRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006504.1 | NP_001006504.1 | 8 - 9 | 1483 | 66 | |
| XM_015288658.2 | XP_015144144.1 | 8 - 9 | 1612 | 66 | ||
| GenBank mRNA | AJ720917.1 | - | 8 - 9 | 1483 | 66 | |
| CR524393.1 | - | 7 - 8 | 1231 | 66 | ||
More Information
Set Membership: |
|

NHLRC2
Genomic Map