Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006510.1 | NP_001006510.1 | 3 - 4 | 425 | 78 | |
| GenBank mRNA | AB185935.1 | - | 3 - 4 | 409 | 78 | |
| AJ720056.1 | - | 3 - 4 | 425 | 78 | ||
More Information
Set Membership: |
|

ZC3H15
Genomic Map