Assay Details
| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006513.1 | NP_001006513.1 | 2 - 3 | 408 | 79 | |
| XM_015289175.2 | XP_015144661.1 | 2 - 3 | 354 | 79 | ||
| GenBank mRNA | AJ719307.1 | - | 2 - 3 | 408 | 79 | |
| BX934894.1 | - | 1 - 2 | 238 | 79 | ||
| FJ655774.1 | - | 1 - 2 | 222 | 79 | ||
More Information
Set Membership: |
|

SLC19A1
Genomic Map