Assay Details
| Gene Symbol: BCS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006520.1 | NP_001006520.1 | 2 - 3 | 438 | 58 | |
| GenBank mRNA | AJ720889.1 | - | 2 - 3 | 438 | 58 | |
More Information
Set Membership: |
|

BCS1L
Genomic Map