Assay Details
| Gene Symbol: DPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006538.1 | NP_001006538.1 | 3 - 4 | 508 | 60 | |
| GenBank mRNA | AJ720102.1 | - | 3 - 4 | 508 | 60 | |
More Information
Set Membership: |
|

DPH2
Genomic Map