Assay Details
| Gene Symbol: TMEM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006541.1 | NP_001006541.1 | 1 - 2 | 220 | 67 | |
| XM_015291021.2 | XP_015146507.1 | 1 - 2 | 201 | 67 | ||
| GenBank mRNA | AJ719736.1 | - | 1 - 2 | 220 | 67 | |
More Information
Set Membership: |
|

TMEM59
Genomic Map