Assay Details
| Gene Symbol: GYG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006558.1 | NP_001006558.1 | 5 - 6 | 673 | 82 | |
| XM_015291687.2 | XP_015147173.1 | 5 - 6 | 679 | 82 | ||
| XM_015291688.2 | XP_015147174.1 | 6 - 7 | 765 | 82 | ||
| XM_015291689.2 | XP_015147175.1 | 6 - 7 | 738 | 82 | ||
| XM_015291691.2 | XP_015147177.1 | 5 - 6 | 702 | 82 | ||
| XM_015291692.2 | XP_015147178.1 | 5 - 6 | 679 | 82 | ||
| XM_025153395.1 | XP_025009163.1 | 5 - 6 | 679 | 82 | ||
| GenBank mRNA | AJ720384.1 | - | 5 - 6 | 673 | 82 | |
More Information
Set Membership: |
|

GYG1
Genomic Map