Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007078.1 | NP_001007079.1 | 2 - 3 | 175 | 59 | |
| GenBank mRNA | AJ621253.1 | - | 2 - 3 | 175 | 59 | |
| EU939770.1 | - | 2 - 3 | 175 | 59 | ||
| GQ421598.1 | - | 2 - 3 | 175 | 59 | ||
| HQ008782.1 | - | 2 - 3 | 175 | 59 | ||
More Information
Set Membership: |
|

CSF2
Genomic Map