Assay Details
| Gene Symbol: ESRRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007081.1 | NP_001007082.1 | 4 - 5 | 670 | 65 | |
| GenBank mRNA | AY702438.1 | - | 4 - 5 | 670 | 65 | |
| BX930078.2 | - | 3 - 4 | 426 | 65 | ||
More Information
Set Membership: |
|

ESRRG
Genomic Map