Assay Details
| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007826.1 | NP_001007827.1 | 1 - 2 | 1059 | 144 | |
| XM_025153838.1 | XP_025009606.1 | 6 - 7 | 4061 | 144 | ||
| GenBank mRNA | AJ720313.1 | - | 1 - 2 | 1059 | 144 | |
More Information
Set Membership: |
|

ABHD17C
Genomic Map