Assay Details
| Gene Symbol: ABHD17C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007826.1 | NP_001007827.1 | 2 - 3 | 1236 | 114 | |
| XM_025153838.1 | XP_025009606.1 | 7 - 8 | 4238 | 114 | ||
| GenBank mRNA | AJ720313.1 | - | 2 - 3 | 1236 | 114 | |
More Information
Set Membership: |
|

ABHD17C
Genomic Map