Assay Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007829.1 | NP_001007830.1 | 2 - 3 | 180 | 101 | |
| GenBank mRNA | AJ720978.1 | - | 2 - 3 | 180 | 101 | |
More Information
Set Membership: |
|

SNAPC5
Genomic Map