Assay Details
| Gene Symbol: RAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007852.1 | NP_001007853.1 | 7 - 8 | 710 | 111 | |
| XM_015281694.2 | XP_015137180.1 | 7 - 8 | 792 | 111 | ||
| GenBank mRNA | AJ719603.1 | - | 7 - 8 | 710 | 111 | |
| CR388910.1 | - | 1 - 2 | 80 | 111 | ||
More Information
Set Membership: |
|

RAP1B
Genomic Map