Assay Details
| Gene Symbol: SLC39A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007933.1 | NP_001007934.1 | 1 - 2 | 481 | 55 | |
| GenBank mRNA | AJ720685.1 | - | 1 - 2 | 481 | 55 | |
| FJ423550.1 | - | 1 - 2 | 101 | 55 | ||
More Information
Set Membership: |
|

SLC39A9
Genomic Map