Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007956.1 | NP_001007957.1 | 3 - 4 | 508 | 96 | |
| GenBank mRNA | AJ719300.1 | - | 3 - 4 | 508 | 96 | |
| CR389193.1 | - | 3 - 4 | 487 | 96 | ||
More Information
Set Membership: |
|

TMED5
Genomic Map