Assay Details
| Gene Symbol: DNAJC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008437.1 | NP_001008437.1 | 5 - 6 | 573 | 75 | |
| XM_015274464.2 | XP_015129950.1 | 5 - 6 | 609 | 75 | ||
| GenBank mRNA | AJ720971.1 | - | 5 - 6 | 573 | 75 | |
More Information
Set Membership: |
|

DNAJC3
Genomic Map