Assay Details
| Gene Symbol: OSBPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008448.1 | NP_001008448.1 | 6 - 7 | 688 | 69 | |
| XM_015296376.2 | XP_015151862.1 | 5 - 6 | 901 | 69 | ||
| XM_015296377.2 | XP_015151863.1 | 6 - 7 | 644 | 69 | ||
| XM_015296379.2 | XP_015151865.1 | 5 - 6 | 791 | 69 | ||
| XM_025142206.1 | XP_024997974.1 | 7 - 8 | 16781 | 69 | ||
| XM_025142207.1 | XP_024997975.1 | 7 - 8 | 16816 | 69 | ||
| GenBank mRNA | AJ719810.1 | - | 6 - 7 | 688 | 69 | |
More Information
Set Membership: |
|

OSBPL2
Genomic Map