Assay Details
| Gene Symbol: GSKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008473.1 | NP_001008473.1 | 2 - 3 | 341 | 88 | |
| XM_015287456.2 | XP_015142942.1 | 2 - 3 | 683 | 88 | ||
| XM_015287457.2 | XP_015142943.1 | 2 - 3 | 414 | 88 | ||
| GenBank mRNA | AJ719458.1 | - | 2 - 3 | 341 | 88 | |
More Information
Set Membership: |
|

GSKIP
Genomic Map